missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Labconco™ Protector™Acid Storage Cabinets
Support Labconco fume hoods and safely store and vent acids and other corrosive liquids
Supplier: Labconco™ LCN9905000
Includes: Vacuum-formed PVC liner tray, two rear vent connection cutouts for venting the cabinet into a fume hood, four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in..
Description
- Corrosion-resistant polyethylene-lined interior
- Manual-closing, non-locking door(s)
Specifications
Acid Storage Cabinet | |
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent | |
24 in. | |
Single Right Hinged Door | |
55.88 cm | |
31.5 to 32.75 in. | |
800 lb. | |
Steel |
22 in. | |
Vacuum formed PVC liner tray, 2 rear vent connection cut outs for venting the cabinet into a fume hood, 4 leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30in. | |
54.43 kg | |
120 lb. | |
362.8 kg | |
ADA | |
80 to 83.18 cm |