Eppendorf™ Protein LoBind™ Tubes
Engineered to minimize sample binding and elevate recovery, Eppendorf™ Protein LoBind™ Tubes safeguard your valuable proteins during processing. With cutting-edge LoBind technology, ensure optimal yields for your Western blotting, mass spectrometry, and protein purification applications.
Supplier:  Eppendorf™ 0030108434
 View more versions of this product
Catalog No. 05414205
Explore available promotions
Description
Description
  • Surface-coating-free, including substances like silicone, plasticizers, biocides or mold release agents to reduce the risk of sample interference.
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
  • Snap, screw, and SnapTec™ cap options available.
Specifications
Specifications
HDPE
0.5 mL
500 Pk.
Non-sterile
Conical
Microcentrifuge Tube (PCR Clean, Protein LoBind)
PCR Clean (Non-sterile)
Colorless
Polypropylene
1 Bag x 500 Tubes
Autoclavable
Snap Cap
18,000 x g