Eppendorf™ Protein LoBind™ Tubes
Engineered to minimize sample binding and elevate recovery, Eppendorf™ Protein LoBind™ Tubes safeguard your valuable proteins during processing. With cutting-edge LoBind technology, ensure optimal yields for your Western blotting, mass spectrometry, and protein purification applications.
$191.68 - $407.00
Specifications
Closure Material HDPE
Material Polypropylene
Sterility Non-sterile
Bottom Shape Conical
Product Type Microcentrifuge Tube (PCR Clean, Protein LoBind)
View More Specs
Products 7
Catalog Number Mfr. No. Closure Type Capacity (Metric) Max. RCF Packaging Price Quantity  
Catalog Number Mfr. No. Closure Type Capacity (Metric) Max. RCF Packaging Price Quantity  
01670413
View Documents
Eppendorf™
0030108531
SnapTec Cap 25 mL 14,000 x g 200 Tubes (5 Bags x 40 Tubes)
Pack of 200 Each for $405.21
01670417
View Documents
Eppendorf™
0030122283
Screw Cap 25 mL 17,000 x g 200 Tubes (4 Bags x 50 Tubes)
Pack of 200 Each for $407.00
05414205
View Documents
Eppendorf™
0030108434
Snap Cap 0.5 mL 18,000 x g 1 Bag x 500 Tubes
Case of 500 Each for $357.24
05414206
View Documents
Eppendorf™
0030108442
Snap Cap 1.5 mL 18,000 x g 1 Bag x 500 Tubes
Case of 500 Each for $357.24
05414207
View Documents
Eppendorf™
0030108450
Snap Cap 2 mL 18,000 x g 1 Bag x 500 Tubes
Case of 500 Each for $357.24
14-282-304
View Documents
Eppendorf™
0030108302
Snap Cap 5 mL 25,000 x g 100 Tubes (2 Bags x 50 Tubes)
Each for $191.68
05-413-917
View Documents
Eppendorf™
30122356
Screw Cap 5 mL 25,000 x g 200 Tubes (2 Bags x 100 Tubes)
Each for $393.13
Description
Description
  • Surface-coating-free, including substances like silicone, plasticizers, biocides or mold release agents to reduce the risk of sample interference.
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
  • Snap, screw, and SnapTec™ cap options available.
Specifications
Specifications
HDPE
Non-sterile
Microcentrifuge Tube (PCR Clean, Protein LoBind)
Polypropylene
Conical
PCR Clean (Non-sterile)
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'