MilliporeSigma™ Supelco™ PTFE Frit, Porosity: 20 μm
Frits are crucial components of the high performance liquid chromatography system design. They act as a depth filter for the concurrent physical removal of small molecules. Medium, 20 μm porosity frits are used for crystalline precipitates and extraction applications.
Supplier: MilliporeSigma™ Supelco™ 57186
Specifications
Crystalline Precipitates, Extraction | |
PTFE | |
20 μm |
3 mL SPE Tubes | |
108 | |
Frit |