missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Supelco™ PTFE Frit, Porosity: 20 μm

Frits are crucial components of the high performance liquid chromatography system design. They act as a depth filter for the concurrent physical removal of small molecules. Medium, 20 μm porosity frits are used for crystalline precipitates and extraction applications.

$792.69 - $792.69

Specifications

For Use With (Application) Crystalline Precipitates, Extraction
Material PTFE
Porosity 20 μm
Type Frit
View More Specs

Products 1
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
111003699
View Documents
MilliporeSigma™ Supelco™
57186
3 mL SPE Tubes
Each for $792.69
 
Description

Description

Specifications

Specifications

Crystalline Precipitates, Extraction
PTFE
20 μm
Frit
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'