PURUS™ Frames for EnviroTack™ Contamination Control Mats
Designed to be used with the PURUS™ EnviroTack™ mat to create a system that is easy to use in any critical area where direct adhesion of mats to a flooring surface is not allowed.
Supplier: PURUS™ PF384701W00
Description
- Easily moved for cleaning
- Non-skid backing ensures frame stays in place
- Fits all PURUS™ EnviroTack™ mats
- Strong and Durable
- Non-Slip made from Polymer Coated polyester textile
- Naturally antimicrobial, mold and mildew resistant
Specifications
965 x 1194 mm | |
Allows for mobility and ease of cleaning | |
Polystyrene |
White | |
3.175 mm | |
914 x 1143 mm PURUS™ Biohybrid™ Mats |