missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyrene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 180831000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pyrene | |
148°C to 152°C | |
393°C | |
Authentic | |
Glass bottle | |
MFCD00004136 | |
05,693 | |
15,8074 | |
Solubility in water: almost insoluble. Other solubilities: soluble in ethanol,ether,benzene and toluene,slightly soluble in carbon tetrachloride | |
C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2 | |
202.25 | |
CHEBI:39106 | |
98% |
129-00-0 | |
Green-Yellow to Yellow | |
210°C | |
97.5% min. (GC) | |
C16H10 | |
100 g | |
04,414 | |
benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene | |
BBEAQIROQSPTKN-UHFFFAOYSA-N | |
pyrene | |
31423 | |
202.25 | |
Crystals and/or Chunks |
Chemical Identifiers
129-00-0 | |
202.25 | |
BBEAQIROQSPTKN-UHFFFAOYSA-N | |
31423 | |
pyrene |
C16H10 | |
MFCD00004136 | |
benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene | |
CHEBI:39106 | |
C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2 |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
GHS Signal Word: Warning
EINECSNumber : 204-927-3
RUO – Research Use Only