missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrene, 98%, Thermo Scientific Chemicals

$50.38 - $150.20

Chemical Identifiers

CAS 129-00-0
Molecular Formula C16H10
Molecular Weight (g/mol) 202.25
MDL Number MFCD00004136
InChI Key BBEAQIROQSPTKN-UHFFFAOYSA-N
Synonym benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene
PubChem CID 31423
ChEBI CHEBI:39106
IUPAC Name pyrene
SMILES C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC180830250
SDS View Documents
Thermo Scientific Chemicals
180830250
25 g Glass bottle
Each for $50.38
 
AC180831000
SDS View Documents
Thermo Scientific Chemicals
180831000
100 g Glass bottle
Each for $150.20
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

129-00-0
202.25
BBEAQIROQSPTKN-UHFFFAOYSA-N
31423
pyrene
C16H10
MFCD00004136
benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene
CHEBI:39106
C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2

Specifications

129-00-0
Green-Yellow to Yellow
210°C
97.5% min. (GC)
C16H10
25 g
04,414
benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene
BBEAQIROQSPTKN-UHFFFAOYSA-N
pyrene
31423
202.25
Crystals and/or Chunks
148°C to 152°C
393°C
Authentic
Glass bottle
MFCD00004136
05,693
15,8074
Solubility in water: almost insoluble. Other solubilities: soluble in ethanol,ether,benzene and toluene,slightly soluble in carbon tetrachloride
C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2
202.25
CHEBI:39106
98%
Pyrene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.

GHS Signal Word: Warning

EINECSNumber : 204-927-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only