Corning™ PYREX™ Class A Buret with Product Standard PTFE Stopcock Plug
Buret has colored markings, fine, sharp lines and large easy-to-read numbers
Supplier:  Corning™ 210310
 View more versions of this product
Catalog No. 03-701-25A
Explore available promotions
Description
Description
Buret has colored markings, fine, sharp lines and large easy-to-read numbers
The capacity tolerance on these burets is established by ASTM E-287 and they are calibrated in accordance with ASTM E-542. Tips are carefully drawn from accurate bore tubing to insure proper drainage rates. This buret has colored markings, fine, sharp lines, and large easy-to-read numbers. The stopcock barrel has a smooth micro-finish to assure leak-resistant performance. Each buret is supplied with a dust cover. All sizes are furnished with a 2 mm bore stopcock plug.
Compliance
Meet ASTM™ E 694, E 542, and E 287
Specifications
Specifications
0.35 in.
10 mL
Glass
56 cm
Dust Cap
PTFE
Class A Burette
ASTM E287, ASTM E542
9 cm
22.04 in.
0.05 mL
1/Cs.
Straight
±0.02 mL
Class A