Corning™ PYREX™ Narrow Mouth Erlenmeyer Culture Flask with Baffles
Usable with shakers—three baffles provide optimal and consistent agitation
$184.38 - $800.81
Specifications
Description Top style: Tooled
Material Borosilicate Glass
For Use With (Application) Ideal for shaker cultures
Neck Style Narrow Mouth
Mouth Style Narrow
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
10-041-5A
Accessories View Documents
Corning™
4450125
125 mL
Case of 6 Each for $652.47
10-041-5B
Accessories View Documents
Corning™
4450250
250 mL
Case of 6 Each for $738.71
10-041-5C
Accessories View Documents
Corning™
4450500
500 mL
Case of 6 Each for $800.81
10-041-5D
View Documents
Corning™
44501L
1 L
Case for $184.38
Description
Description
Heavy tooled rim for maximum durability. The same baffle design as found on our Delong flasks promotes optimal and consistent solution agitation when used with flask shakers.
Specifications
Specifications
Top style: Tooled
Ideal for shaker cultures
Narrow
Baffled
Borosilicate Glass
Narrow Mouth
Non-sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'