Corning™ PYREX™ Low Form Class B Lifetime Red™ Cylinders with Single Metric Scale, Double Pour Out
Calibrated To Contain
Supplier:  Corning™ 3044100
 View more versions of this product
Catalog No. 08-562-10B
Explore available promotions
Description
Description
Calibrated 'to contain.' The scale and figures are durable white enamel which stand out clearly against the red color-band on these Lifetime Red cylinders. The cylinder is a Tuttle type of low form for stability and features a double pourout for convenience.
Specifications
Specifications
Round
Graduated Cylinder
To Contain
White
Hexagonal
157 mm
1.5 in.
100 mL
Lifetime red background
2 mL
2 mL
±1.2 mL
6.1 in.
Non-sterile
ASTM E438
Lifetime Red
Borosilicate Glass
39 mm
Provides Clear, Accurate Measurements for Delivering or Containing Liquids
Single
Autoclavable
Videos
SDS
Documents
Documents
Accessories