Corning™ PYREX™ Low Form Class B Lifetime Red™ Cylinders with Single Metric Scale, Double Pour Out
Calibrated To Contain
$148.21 - $1,540.45
Specifications
Base Type Hexagonal
Calibration To Contain
Certifications/Compliance ASTM E438
Class Lifetime Red
Type Graduated Cylinder
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity  
08-562-10B
Accessories View Documents
Corning™
3044100
100 mL 2 mL N/A
08-562-10A
Accessories View Documents
Corning™
304450
50 mL 1 mL N/A
Description
Description
Calibrated 'to contain.' The scale and figures are durable white enamel which stand out clearly against the red color-band on these Lifetime Red cylinders. The cylinder is a Tuttle type of low form for stability and features a double pourout for convenience.
Specifications
Specifications
Hexagonal
ASTM E438
Graduated Cylinder
Round
Borosilicate Glass
Lifetime red background
Autoclavable
To Contain
Lifetime Red
White
Non-sterile
Provides Clear, Accurate Measurements for Delivering or Containing Liquids
Single
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'