Corning™ PYREX™ Cylindrical Tanks
Tops are ground to close tolerances to ensure tight cover fits
Supplier:  Corning™ 694513L
 View more versions of this product
Catalog No. 05-719-8D
Explore available promotions
Description
Description
A general-purpose jar with tops ground to close tolerances to insure a tight cover fit. The 152 mm jar edge is flat within 0.1 mm; all others are flat within 0.25 mm. Covers are not supplied. Do not use with heat, pressure, or vacuum applications.
Specifications
Specifications
3.5 gal.
Ground
Cylindrical Chromatography Tank
0.25 mm
45.7 cm
22.2 cm
13.2 L
Pyrex Glass
Cylindrical
18 in.
8.75 in.
Product Suggestions
Product Suggestions