Learn More
Corning™ PYREX™ Cylindrical Tanks
Tops are ground to close tolerances to ensure tight cover fits
$1,057.28 - $6,365.39
Specifications
Closure Type Ground
Material Pyrex Glass
Type Cylindrical Chromatography Tank
Shape Cylindrical
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (English) Diameter (English) Outer Height (English) Tolerance Price Quantity  
Catalog Number Mfr. No. Capacity (English) Diameter (English) Outer Height (English) Tolerance Price Quantity  
05-719-8B
View Documents
Corning™
69456L
1.7 gal. 6 in. 18 in. 0.1 mm N/A
Only null left
05-719-8D
View Documents
Corning™
694513L
3.5 gal. 8.75 in. 18 in. 0.25 mm N/A
Only null left
05-719-8F
View Documents
Corning™
694536L
9.5 gal. 12 in. 24 in. 0.25 mm N/A
Only null left
Description
Description
A general-purpose jar with tops ground to close tolerances to insure a tight cover fit. The 152 mm jar edge is flat within 0.1 mm; all others are flat within 0.25 mm. Covers are not supplied. Do not use with heat, pressure, or vacuum applications.
Specifications
Specifications
Ground
Pyrex Glass
Cylindrical Chromatography Tank
Cylindrical
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'