Corning™ PYREX™ Dispensing Burets with Straight Bore Product Standard PTFE Stopcock
For dispensing large volumes of liquids rapidly and accurately
Supplier:  Corning™ 2094500
 View more versions of this product
Catalog No. 03-693-1B
Explore available promotions
Description
Description
Used when dispensing large volumes of liquids rapidly and accurately. Made from close tolerance, accurate bore tubing with uniform walls, these burets have a permanently colored scale. Tips are carefully drawn and ground to reduce chipping. Provided with a PTFE stopcock plug to reduce freezing and eliminate lubricant contamination. The smooth micro-finish of the stopcock barrel assures a precision fit and optimum leak-resistant performance.
Specifications
Specifications
PTFE
41 mm
26.5 in.
5 mL
675 mm
1/Cs.
4
±2.5 mL
1.6 in.
500 mL
Glass
Dispensing Large Volumes of Liquids Rapidly and Accurately
1/Pk.
Straight Bore Product Standard PTFE Stopcock
PTFE