Corning™ PYREX™ Dispensing Burets with Straight Bore Product Standard PTFE Stopcock
For dispensing large volumes of liquids rapidly and accurately
$388.28 - $614.17
Specifications
Closure Material | PTFE |
---|---|
Material | Glass |
For Use With (Application) | Dispensing Large Volumes of Liquids Rapidly and Accurately |
No. per Pack | 1/Pk. |
No. per Case | 1/Cs. |
Description
Used when dispensing large volumes of liquids rapidly and accurately. Made from close tolerance, accurate bore tubing with uniform walls, these burets have a permanently colored scale. Tips are carefully drawn and ground to reduce chipping. Provided with a PTFE stopcock plug to reduce freezing and eliminate lubricant contamination. The smooth micro-finish of the stopcock barrel assures a precision fit and optimum leak-resistant performance.
Specifications
PTFE | |
Dispensing Large Volumes of Liquids Rapidly and Accurately | |
1/Cs. | |
PTFE |
Glass | |
1/Pk. | |
Straight Bore Product Standard PTFE Stopcock |