missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ PYREX™ Long Neck Flat Bottom Flasks, Standard Taper Joint

Long Necks

Supplier:  Corning™ 4080500

 View more versions of this product

Catalog No. 10-044B


Explore available promotions

Description

Description

With full length outer standard taper joints.

Specifications

Specifications

4 in.
Borosilicate Glass
19.5 cm
12/Cs.
Reusable
103 mm
500 mL
7.6 in.
Long
24/40
Flat
Videos
SDS
Documents

Documents

Promotions

Promotions