Corning™ PYREX™ Long Neck Flat Bottom Flasks, Standard Taper Joint
Long Necks
$1228.49 - $1458.87
Specifications
Material Borosilicate Glass
Neck Style Long
No. per Case 12/Cs.
Standard Taper Joint 24/40
Disposable Reusable
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
10-044A
View Documents
Corning™
4080250
250 mL
Case of 12 Each for $1,228.49
10-044B
View Documents
Corning™
4080500
500 mL
Case of 12 Each for $1,458.87
Description
Description
With full length outer standard taper joints.
Specifications
Specifications
Borosilicate Glass
12/Cs.
Reusable
Long
24/40
Flat
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'