Corning™ PYREX™ Gas Dispersion Tubes With Fritted Cylinders
Used to disperse or scrub gas in liquids.
Supplier: Corning™ 3953312C
Description
These immersion tubes have a 30 mm diameter medium porosity (10 μm to 15 μm) fritted disc and are particularly useful for drawing off the supernatant liquid from a precipitate. They are made from Code 7740 borosilicate glass. Any precipitate which might be disturbed will be held on the filter. Solid chemicals, made in a beaker or flask, can be filtered and washed without being transferred.
Specifications
Pyrex Gas Dispersion Tube | |
40 to 60 μm | |
0.31 in. | |
9.8 in. | |
Used to disperse or scrub gas in liquids |
Coarse porosity fritted cylinder | |
Coarse | |
8 mm | |
250 mm | |
Test tubes or flasks |