Corning™ PYREX™ Media Bottle High-Temperature Caps, Open-Top
With drip-free pouring ring
Supplier:  Corning™ 139532HTSC
 View more versions of this product
Catalog No. 06-414-10
Explore available promotions
Description
Description
These are various sizes and types of screw caps for use with storage bottles.
Specifications
Specifications
Red
PBT
10/Pk.
GL 32
Open Top Cap
PBT
24.2 mm
10/Cs.
Non-sterile
Red
Storage bottle
High Temperature