missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ PYREX™ Media Bottle High-Temperature Caps, Open-Top

With drip-free pouring ring

$157.15 - $263.41

Specifications

Color Red
Material PBT
No. per Case 10/Cs.
No. per Pack 10/Pk.
Sterility Non-sterile
View More Specs

Products 3
Catalog Number Mfr. No. Thread Size (Metric) Price Quantity  
Catalog Number Mfr. No. Thread Size (Metric) Price Quantity  
06-414-11
View Documents
Corning™
139525HTSC
GL 25
Case of 10 Each for $157.15
 
06-414-10
View Documents
Corning™
139532HTSC
GL 32
Case of 10 Each for $230.05
 
06-414-9
Accessories View Documents
Corning™
139545HTSC
GL 45
Case of 10 Each for $263.41
 
Description

Description

These are various sizes and types of screw caps for use with storage bottles.

Specifications

Specifications

Red
10/Cs.
Non-sterile
Open Top Cap
PBT
PBT
10/Pk.
Red
Storage bottle
High Temperature
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'