Corning™ PYREX™ Media Bottle Septa
For use with open-top PBT caps
Supplier: Corning™ 139545TS
Description
These are various sizes and types of screw caps for use with storage bottles.
Specifications
Natural | |
10/Cs. | |
10 Cs. | |
GL45 open top PBT screw cap | |
No Closure |
PTFE-faced Silicone Rubber | |
10/Pk. | |
Non-sterile | |
GL 45 |