Corning™ PYREX™ Reusable Test Tubes with Beaded Rim
Sizes from 10 to 20mm O.D. have marking spot
Supplier:  Corning™ 980010
 View more versions of this product
Catalog No. 14-955A
Description
Description
These tubes are made from special tubing to give the optimum wall thickness. They are well annealed, resistant to heat, and chemically stable. Rims are fire-polished.
Specifications
Specifications
Plain
75 mm
10 mm
3 mL
Borosilicate Glass