Corning™ PYREX™ Round-Bottom Glass Centrifuge Tubes
Without graduations
Supplier:  Corning™ 842250
 View more versions of this product
Catalog No. 05-558-5B
Explore available promotions
Description
Description
These PYREX™ round bottom, ungraduated centrifuge tubes have a pourout spout for easy decanting.
Specifications
Specifications
Phenolic
Reusable
Useful for the handling of sputum specimens, digestion, shaking, neutralizing and centrifuging
12
24-410
Cylindrical
Yes
Centrifuge Tube
50 mL
Borosilicate Glass
Rubber
12
Non-sterile
122 mm
29 mm
Videos
SDS
Documents
Documents
Accessories