Corning™ PYREX™ Solid Glass Beads for Distillation Columns
Available in four sizes
Supplier:  Corning™ C72684
 View more versions of this product
Catalog No. 11-312-10B
Explore available promotions
Description
Description
Available in four sizes
Useful as packing for distillation columns, mixing beads, and boiling stones. Beads are packaged in 0.45 kg. (1 lb.) packs, which have a packing volume of approximately 360 cm3 or 22 cubic inches.
Order Info
Pack size is 1 lb.
Specifications
Specifications
Glass
4 mm