Corning™ PYREX™ Solid Glass Beads for Distillation Columns
Available in four sizes
$2167.28 - $3313.08
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity | |||||
11-312-10A
|
Corning™
72683 |
3 mm |
Case for $3,313.08
|
|
|||||
11-312-10C
|
Corning™
72685 |
5 mm |
Case for $2,167.28
|
|
|||||
11-312-10B
|
Corning™
C72684 |
4 mm |
Case for $2,802.99
|
|
|||||
11-312-10D
|
Corning™
72686 |
6 mm |
Case for $2,533.26
|
|
|||||
Description
Available in four sizes
Useful as packing for distillation columns, mixing beads, and boiling stones. Beads are packaged in 0.45 kg. (1 lb.) packs, which have a packing volume of approximately 360 cm3 or 22 cubic inches.
Order Info
Pack size is 1 lb.
Specifications
Glass |