Corning™ PYREX™ VISTA™ Rimless Reusable Glass Culture Tubes
Functionality, economy, and quality
Supplier:  Corning™ 708206
 View more versions of this product
Catalog No. 07-250-132
Explore available promotions
Description
Description
These test tubes are annealed, resistant to heat and chemically stable. Rims are fire-polished. The 25 mm diameter tubes do not have a marking spot.
Specifications
Specifications
Fire Polished
50 mm
Reusable
Blue Marking Spot
Non-sterile
6 mm
0.5 mL
Borosilicate Glass
For convenient plugging and rack storage
No
No
Culture Tube