Corning™ PYREX™ VISTA™ Rimless Reusable Glass Culture Tubes
Functionality, economy, and quality
$83.15 - $1400.00
Specifications
Design Type Fire Polished
Material Borosilicate Glass
Disposable Reusable
For Use With For convenient plugging and rack storage
Rimmed No
View More Specs
Products 10
Catalog Number Mfr. No. Capacity (Metric) Length (Metric) Diameter (Metric) Label Type Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Length (Metric) Diameter (Metric) Label Type Price Quantity  
07-250-122
View Documents
Corning™
7082012
5 mL 75 mm 12 mm Blue Marking Spot
Pack of 50 Each for $87.97
Case of 4 Pack for $351.88
07-250-124
View Documents
Corning™
7082016
11 mL 100 mm 16 mm Blue Marking Spot
Pack of 50 Each for $109.79
Case of 4 Pack for $439.16
07-250-121
View Documents
Corning™
7082010
3 mL 75 mm 10 mm Blue Marking Spot
Pack of 50 Each for $83.15
Case of 4 Pack for $332.60
07-250-126
View Documents
Corning™
C7082016XX
20 mL 150 mm 16 mm Blue Marking Spot
Case of 200 Each for $339.32
07-250-127
View Documents
Corning™
7082018
27 mL 150 mm 18 mm Blue Marking Spot
Case of 200 Each for $391.32
07-250-128
View Documents
Corning™
7082020
34 mL 150 mm 20 mm Blue Marking Spot
Pack of 50 Each for $137.40
Case of 4 Pack for $549.60
07-250-129
View Documents
Corning™
7082022
50 mL 175 mm 22 mm None
Pack of 50 Each for $246.96
Case of 4 Pack for $987.84
07-250-132
View Documents
Corning™
708206
0.5 mL 50 mm 6 mm Blue Marking Spot
Case of 50 Each for $343.57
07-250-130
View Documents
Corning™
7082025
55 mL 150 mm 25 mm None
Pack of 50 Each for $304.20
Case of 4 Pack for $1,216.80
07-250-131
View Documents
Corning™
7082025X
70 mL 200 mm 25 mm None
Case of 50 Each for $1,400.00
Description
Description
These test tubes are annealed, resistant to heat and chemically stable. Rims are fire-polished. The 25 mm diameter tubes do not have a marking spot.
Specifications
Specifications
Fire Polished
Reusable
No
No
Borosilicate Glass
For convenient plugging and rack storage
Non-sterile
Culture Tube
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'