Corning™ PYREX™ Watch Glass/Beaker Cover with Fire-Polished Edges
Molded for table stacking
Supplier:  Corning™ 9985150
 View more versions of this product
Catalog No. 02-612F
Explore available promotions
Description
Description
Molded for table stacking
These watch glasses possess high chemical durability, thus preventing contamination of solutions. They are molded to the same radius of curvature for convenient stacking. The heavy wall and ­uniform fire-polished edge provide mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected to occur.
Specifications
Specifications
Reusable
Round
Used as Beaker Covers
Polished
12/Pk.
15 cm
Borosilicate Glass
Watch Glass
For 2000 mL beaker
72/Cs.
5.9 in.