Corning™ PYREX™ Watch Glass/Beaker Cover with Fire-Polished Edges
Molded for table stacking
$103.69 - $1446.18
Specifications
Disposable Reusable
Material Borosilicate Glass
Shape Round
Type Watch Glass
For Use With (Application) Used as Beaker Covers
View More Specs
Products 6
Catalog Number Mfr. No. Diameter (Metric) For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) For Use With (Equipment) Price Quantity  
02-612E
View Documents Promotion Details
Corning™
9985125
12.5 cm For 1000 mL beaker
Pack of 12 Each for $186.54
Case of 6 Pack for $1,119.24
02-612B
View Documents
Corning™
998575
7.5 cm For 250 mL beaker
Pack of 12 Each for $112.08
Case of 12 Pack for $1,344.96
02-612A
View Documents
Corning™
998565
6.5 cm For 150 mL beaker
Pack of 12 Each for $103.69
Case of 12 Pack for $1,244.28
02-612F
View Documents Promotion Details
Corning™
9985150
15 cm For 2000 mL beaker
Pack of 12 Each for $262.21
Case of 6 Pack for $1,573.26
02-612D
View Documents
Corning™
9985100
10 cm For 600 mL beaker
Pack of 12 Each for $132.00
Case of 12 Pack for $1,584.00
02-612C
View Documents Promotion Details
Corning™
998590
9 cm For 400 mL beaker
Case of 144 Each for $1,446.18
Description
Description
Molded for table stacking
These watch glasses possess high chemical durability, thus preventing contamination of solutions. They are molded to the same radius of curvature for convenient stacking. The heavy wall and ­uniform fire-polished edge provide mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected to occur.
Specifications
Specifications
Reusable
Round
Used as Beaker Covers
12/Pk.
Borosilicate Glass
Watch Glass
Polished
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'