missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyruvic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1387536

 View more versions of this product

Catalog No. AAA1387536

Explore available promotions

Description

Description

Pyruvic acid is used as a component in culture broths and media as commercial red seaweed polysaccharide. It is involved in the construction of amino acid alanine as well as supplies energy to living cells via citric acid cycle (Krebs cycle). It reacts with N-acetyl mannosamine by an aldol-type condensation to prepare sialic acid. It is employed to study the cultivation of soil bacteria as micro colonies using soil substrate membrane system. It finds application in liquid chromatography and in the determination of organic acids in red wine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyruvic acid is used as a component in culture broths and media as commercial red seaweed polysaccharide. It is involved in the construction of amino acid alanine as well as supplies energy to living cells via citric acid cycle (Krebs cycle). It reacts with N-acetyl mannosamine by an aldol-type condensation to prepare sialic acid. It is employed to study the cultivation of soil bacteria as micro colonies using soil substrate membrane system. It finds application in liquid chromatography and in the determination of organic acids in red wine.

Solubility
Miscible with chloroform and methanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong bases and reducing agents.
Specifications

Specifications

Pyruvic acid
11°C to 12°C
1.2
83°C (181°F)
Pungent
1.4295
500 g
506211
pyruvic acid, pyroracemic acid, 2-oxopropionic acid, acetylformic acid, alpha-ketopropionic acid, 2-ketopropionic acid, 2-oxopropanoate, propanoic acid, 2-oxo, 2-oxo-propionic acid, 2-oxopropansaeure
LCTONWCANYUPML-UHFFFAOYSA-N
2-oxopropanoic acid
1060
88.06
Liquid
127-17-3
1.267
164°C to 166°C
98%
C3H4O3
MFCD00002585
UN3265
14,8021
Miscible with chloroform and methanol.
CC(=O)C(O)=O
88.06
CHEBI:32816
98%

Chemical Identifiers

127-17-3
88.06
LCTONWCANYUPML-UHFFFAOYSA-N
1060
2-oxopropanoic acid
C3H4O3
MFCD00002585
pyruvic acid, pyroracemic acid, 2-oxopropionic acid, acetylformic acid, alpha-ketopropionic acid, 2-ketopropionic acid, 2-oxopropanoate, propanoic acid, 2-oxo, 2-oxo-propionic acid, 2-oxopropansaeure
CHEBI:32816
CC(=O)C(O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 204-824-3

RTECSNumber : UZ0829800

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only