Learn More
Pyruvic acid, 98%, Thermo Scientific Chemicals
$125.26 - $1477.38
Chemical Identifiers
CAS | 127-17-3 |
---|---|
Molecular Formula | C3H4O3 |
Molecular Weight (g/mol) | 88.06 |
MDL Number | MFCD00002585 |
InChI Key | LCTONWCANYUPML-UHFFFAOYSA-N |
Synonym | pyruvic acid, pyroracemic acid, 2-oxopropionic acid, acetylformic acid, alpha-ketopropionic acid, 2-ketopropionic acid, 2-oxopropanoate, propanoic acid, 2-oxo, 2-oxo-propionic acid, 2-oxopropansaeure |
PubChem CID | 1060 |
ChEBI | CHEBI:32816 |
IUPAC Name | 2-oxopropanoic acid |
SMILES | CC(=O)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1387522
|
Thermo Scientific Chemicals
A1387522 |
100 g |
Each for $125.26
|
|
||||||
AAA1387536
|
Thermo Scientific Chemicals
A1387536 |
500 g |
Each for $396.27
|
|
||||||
AAA138750E
|
Thermo Scientific Chemicals
A138750E |
2500 g |
Each for $1,477.38
|
|
||||||
Description
Pyruvic acid is used as a component in culture broths and media as commercial red seaweed polysaccharide. It is involved in the construction of amino acid alanine as well as supplies energy to living cells via citric acid cycle (Krebs cycle). It reacts with N-acetyl mannosamine by an aldol-type condensation to prepare sialic acid. It is employed to study the cultivation of soil bacteria as micro colonies using soil substrate membrane system. It finds application in liquid chromatography and in the determination of organic acids in red wine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyruvic acid is used as a component in culture broths and media as commercial red seaweed polysaccharide. It is involved in the construction of amino acid alanine as well as supplies energy to living cells via citric acid cycle (Krebs cycle). It reacts with N-acetyl mannosamine by an aldol-type condensation to prepare sialic acid. It is employed to study the cultivation of soil bacteria as micro colonies using soil substrate membrane system. It finds application in liquid chromatography and in the determination of organic acids in red wine.
Solubility
Miscible with chloroform and methanol.
Notes
Store in cool place. Incompatible with strong oxidizing agents, strong bases and reducing agents.
Chemical Identifiers
127-17-3 | |
88.06 | |
LCTONWCANYUPML-UHFFFAOYSA-N | |
1060 | |
2-oxopropanoic acid |
C3H4O3 | |
MFCD00002585 | |
pyruvic acid, pyroracemic acid, 2-oxopropionic acid, acetylformic acid, alpha-ketopropionic acid, 2-ketopropionic acid, 2-oxopropanoate, propanoic acid, 2-oxo, 2-oxo-propionic acid, 2-oxopropansaeure | |
CHEBI:32816 | |
CC(=O)C(O)=O |
Specifications
127-17-3 | |
1.267 | |
83°C (181°F) | |
C3H4O3 | |
MFCD00002585 | |
UN3265 | |
14,8021 | |
Miscible with chloroform and methanol. | |
CC(=O)C(O)=O | |
88.06 | |
CHEBI:32816 | |
98% | |
Pyruvic Acid |
11°C to 12°C | |
164°C to 166°C | |
98% | |
1.4295 | |
100 g | |
506211 | |
pyruvic acid, pyroracemic acid, 2-oxopropionic acid, acetylformic acid, alpha-ketopropionic acid, 2-ketopropionic acid, 2-oxopropanoate, propanoic acid, 2-oxo, 2-oxo-propionic acid, 2-oxopropansaeure | |
LCTONWCANYUPML-UHFFFAOYSA-N | |
2-oxopropanoic acid | |
1060 | |
88.06 | |
Liquid |
Safety and Handling
GHS H Statement
H314-H318-H227
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 204-824-3
RTECSNumber : UZ0829800
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only