missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ pZerve Cryopreservation Solution

A cryopreservation solution that does not contain dimethyl sulfoxide (DMSO) and serum.

Supplier:  MP Biomedicals™ 092030346

 View more versions of this product

Catalog No. MP92030346


Explore available promotions

Description

Description

pZerve has been successfully used with a variety of cell types including mast cells, dendritic cells, and mesenchymal cells. It can be used for cells cultured in serum-free medium or with serum-containing medium.

Specifications

Specifications

Store at 2° to 8°C
Solution
Cryopreservation
pZerve Cryopreservation Media
Cryopreservation Media
20 mL
Videos
SDS
Documents

Documents

Promotions

Promotions