missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Quercetin hydrate, 95%, Thermo Scientific Chemicals
$49.77 - $215.40
Chemical Identifiers
CAS | 849061-97-8 |
---|---|
Molecular Formula | C15H10O7 |
Molecular Weight (g/mol) | 302.24 |
MDL Number | MFCD03847906 |
InChI Key | REFJWTPEDVJJIY-UHFFFAOYSA-N |
Synonym | quercetin hydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, quercetinhydrate, quercetine sophoretin /, regid_for_cid_747934, regid_for_cid_16212154, 3,3;,4;,5,7-pentahydroxyflavon, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxychromen-4-one, hydrate |
PubChem CID | 16212154 |
IUPAC Name | 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate |
SMILES | OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C=C2)=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC174070100
|
Thermo Scientific Chemicals
174070100 |
10 g | Glass bottle |
Each for $49.77
|
|
||||
AC174070250
|
Thermo Scientific Chemicals
174070250 |
25 g | Glass bottle |
Each for $88.10
|
|
||||
AC174071000
|
Thermo Scientific Chemicals
174071000 |
100 g | Glass bottle |
Each for $215.40
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
849061-97-8 | |
302.24 | |
REFJWTPEDVJJIY-UHFFFAOYSA-N | |
16212154 | |
OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C=C2)=C1 |
C15H10O7 | |
MFCD03847906 | |
quercetin hydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, quercetinhydrate, quercetine sophoretin /, regid_for_cid_747934, regid_for_cid_16212154, 3,3;,4;,5,7-pentahydroxyflavon, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxychromen-4-one, hydrate | |
2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate |
Specifications
849061-97-8 | |
Yellow | |
94% min. (HPLC) | |
C15H10O7 | |
10 g | |
14, 8034 | |
Solubility in water: practically insoluble. Other solubilities: soluble in ethanol,acetone,pyridine,,glacial acetic acid | |
OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C=C2)=C1 | |
302.24 | |
302.24 | |
Crystalline Powder |
>300°C | |
Authentic | |
Glass bottle | |
MFCD03847906 | |
18, 242 | |
quercetin hydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, quercetinhydrate, quercetine sophoretin /, regid_for_cid_747934, regid_for_cid_16212154, 3,3;,4;,5,7-pentahydroxyflavon, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxychromen-4-one, hydrate | |
REFJWTPEDVJJIY-UHFFFAOYSA-N | |
2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate | |
16212154 | |
95% | |
Quercetin hydrate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Danger
RTECSNumber : LK8950000
RUO – Research Use Only