Dynalon™ Quick Mist™ HDPE Sprayer Bottles
QuickMist reusable hand sprayer bottle. Trigger-type operation to provide fine mist with large pattern. Trigger assembly chemically resistant polypropylene with plastic strainer. Easily cleaned valve and intake. Excellent for spraying all types of thin liquids. Not suitable for use with food products.
Supplier:  Dynalon™ 605154EMD
 View more versions of this product
Catalog No. S37064
Explore available promotions
Description
Description
  • Excellent for spraying all types of thick liquids
  • Fine mist with large pattern
  • Not suitable for use with food products
Specifications
Specifications
1000 mL, 32 oz.
3.4375 x 9.39 in.
Translucent
0.20 lb.
No
Polypropylene
QuickMist™ Spray Bottle
HDPE
0.0907kg
Yes
Cylindrical
Yes