Quidel QuickVue+™ One-Step hCG Combo Lateral Flow Test Kit
10mIU/mL sensitivity in serum; 20mIU/mL sensitivity in urine
Supplier: Quidel Q00179
Includes: Kits contain test devices, disposable dropper, package insert, and procedure card
Description
- CPT Codes: 81025 (urine), 84703 (serum)
- CLIA Complexity: Waived (urine), Non-Waived (serum)
- Test results within five minutes (serum); three minutes (urine)
- Two-color plus/minus (±) endpoint for easy-to-read and interpret results
- Immunoassay for the qualitative detection of hCG in serum or urine
- >99% sensitivity, specificity, and accuracy
- For in vitro diagnostic use
Specifications
>99% | |
Waived, Non-waived | |
Serum, Urine | |
Test device, disposable dropper, package insert and procedure card | |
3 min. for urine, 5 min. for serum |
81025, 84703 | |
hCG | |
hCG Urine/Serum Test Kit | |
90 Tests |