missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Quidel QuickVue+™ One-Step hCG Combo Lateral Flow Test Kit
10mIU/mL sensitivity in serum; 20mIU/mL sensitivity in urine
$133.29 - $327.42
Specifications
Accuracy | >99% |
---|---|
CPT Code | 81025, 84703 |
Clia Complexity | Waived, Non-waived |
Detectable Analytes | hCG |
Sample Type | Serum, Urine |
Includes: Kits contain test devices, disposable dropper, package insert, and procedure card
Catalog Number | Mfr. No. | Quantity | Type | Sample Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Sample Type | Price | Quantity | |||
23043065
|
Quidel
Q00179 |
90 Tests | hCG Urine/Serum Test Kit | Serum, Urine |
Pack of 90 Each for $327.42
|
|
|||
23043060
|
Quidel
Q00178 |
30 Tests | hCG Urine/Serum Test Kit | Serum, Urine |
Pack of 30 Each for $133.29
|
|
|||
Description
- CPT Codes: 81025 (urine), 84703 (serum)
- CLIA Complexity: Waived (urine), Non-Waived (serum)
- Test results within five minutes (serum); three minutes (urine)
- Two-color plus/minus (±) endpoint for easy-to-read and interpret results
- Immunoassay for the qualitative detection of hCG in serum or urine
- >99% sensitivity, specificity, and accuracy
- For in vitro diagnostic use
Specifications
>99% | |
Waived, Non-waived | |
Serum, Urine | |
Test device, disposable dropper, package insert and procedure card |
81025, 84703 | |
hCG | |
hCG Urine/Serum Test Kit | |
3 min. for urine, 5 min. for serum |