missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Quinaldine Red 95.0+%, TCI America™
Supplier: TCI America Q000725G
Specifications
Quinaldine Red | |
C21H23IN2 | |
JOLANDVPGMEGLK-UHFFFAOYSA-M | |
2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide | |
5702759 | |
≥95.0% (T) | |
25 g |
117-92-0 | |
MFCD00011968 | |
[I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C | |
430.33 | |
430.33 | |
Black/Green | |
Crystal/Powder |
Chemical Identifiers
117-92-0 | |
430.33 | |
JOLANDVPGMEGLK-UHFFFAOYSA-M | |
2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide |
C21H23IN2 | |
MFCD00011968 | |
5702759 | |
[I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C |
Safety and Handling
TSCA : Yes