missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(+)-1-(4-Bromophenyl)ethylamine hydrochloride, 97%, 99+% ee, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 367170050

 View more versions of this product

Catalog No. AC367170050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(R)-(+)-1-(4-Bromophenyl)ethylamine hydrochloride
64265-77-6
100.0
Authentic
Glass bottle
MFCD02259386
r-1-4-bromophenyl ethanamine hydrochloride, r-+-1-4-bromophenyl ethylamine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride, 1r-1-4-bromophenyl ethylamine, chloride, r-1-4-bromophenyl ethanamine hcl, r-1-4-bromophenyl ethylamine hcl, r-1-4-bromophenyl ethanaminehydrochloride, r-1-4-bromophenyl ethylaminehydrochloride, 1r-1-4-bromophenyl ethan-1-amine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride 1:1
BQCAANUXMMQVAY-UHFFFAOYNA-N
(1R)-1-(4-bromophenyl)ethanamine;hydrochloride
2734642
97%,99+% ee
≥99%
96.0
99.0% min. (HPLC)
96% min. (HPLC)
C8H11BrClN
5g
Solubility in water: soluble in water. Other solubilities: miscible with alcohols
Cl.CC(N)C1=CC=C(Br)C=C1
236.54
236.54

Chemical Identifiers

64265-77-6
236.54
BQCAANUXMMQVAY-UHFFFAOYNA-N
2734642
Cl.CC(N)C1=CC=C(Br)C=C1
C8H11BrClN
MFCD02259386
r-1-4-bromophenyl ethanamine hydrochloride, r-+-1-4-bromophenyl ethylamine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride, 1r-1-4-bromophenyl ethylamine, chloride, r-1-4-bromophenyl ethanamine hcl, r-1-4-bromophenyl ethylamine hcl, r-1-4-bromophenyl ethanaminehydrochloride, r-1-4-bromophenyl ethylaminehydrochloride, 1r-1-4-bromophenyl ethan-1-amine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride 1:1
(1R)-1-(4-bromophenyl)ethanamine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only