Learn More
(R)-(+)-1-(4-Bromophenyl)ethylamine hydrochloride, 97%, 99+% ee, Thermo Scientific™
Chemical Identifiers
CAS | 64265-77-6 |
---|---|
Molecular Formula | C8H11BrClN |
Molecular Weight (g/mol) | 236.54 |
MDL Number | MFCD02259386 |
InChI Key | BQCAANUXMMQVAY-UHFFFAOYNA-N |
Synonym | r-1-4-bromophenyl ethanamine hydrochloride, r-+-1-4-bromophenyl ethylamine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride, 1r-1-4-bromophenyl ethylamine, chloride, r-1-4-bromophenyl ethanamine hcl, r-1-4-bromophenyl ethylamine hcl, r-1-4-bromophenyl ethanaminehydrochloride, r-1-4-bromophenyl ethylaminehydrochloride, 1r-1-4-bromophenyl ethan-1-amine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride 1:1 |
PubChem CID | 2734642 |
IUPAC Name | (1R)-1-(4-bromophenyl)ethanamine;hydrochloride |
SMILES | Cl.CC(N)C1=CC=C(Br)C=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC367170010
|
Thermo Scientific Chemicals
367170010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC367170050
|
Thermo Scientific Chemicals
367170050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
64265-77-6 | |
236.54 | |
BQCAANUXMMQVAY-UHFFFAOYNA-N | |
2734642 | |
Cl.CC(N)C1=CC=C(Br)C=C1 |
C8H11BrClN | |
MFCD02259386 | |
r-1-4-bromophenyl ethanamine hydrochloride, r-+-1-4-bromophenyl ethylamine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride, 1r-1-4-bromophenyl ethylamine, chloride, r-1-4-bromophenyl ethanamine hcl, r-1-4-bromophenyl ethylamine hcl, r-1-4-bromophenyl ethanaminehydrochloride, r-1-4-bromophenyl ethylaminehydrochloride, 1r-1-4-bromophenyl ethan-1-amine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride 1:1 | |
(1R)-1-(4-bromophenyl)ethanamine;hydrochloride |
Specifications
64265-77-6 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD02259386 | |
r-1-4-bromophenyl ethanamine hydrochloride, r-+-1-4-bromophenyl ethylamine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride, 1r-1-4-bromophenyl ethylamine, chloride, r-1-4-bromophenyl ethanamine hcl, r-1-4-bromophenyl ethylamine hcl, r-1-4-bromophenyl ethanaminehydrochloride, r-1-4-bromophenyl ethylaminehydrochloride, 1r-1-4-bromophenyl ethan-1-amine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride 1:1 | |
BQCAANUXMMQVAY-UHFFFAOYNA-N | |
(1R)-1-(4-bromophenyl)ethanamine;hydrochloride | |
2734642 | |
97%,99+% ee |
96.0 | |
99.0% min. (HPLC) | |
96% min. (HPLC) | |
C8H11BrClN | |
1g | |
Solubility in water: soluble in water. Other solubilities: miscible with alcohols | |
Cl.CC(N)C1=CC=C(Br)C=C1 | |
236.54 | |
236.54 | |
(R)-(+)-1-(4-Bromophenyl)ethylamine hydrochloride, ≥99% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only