Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
Supplier:  Thermo Scientific™ 4000294
 View more versions of this product
Catalog No. 15-467-421
Explore available promotions
Description
Description
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
  • Warranty varies by country, please contact us for details
Specifications
Specifications
Silver
All CryoMed controlled-rate freezers
Straw Rack
40 0.25/0.5 mL Straws
40 0.25/0.5 mL Straws