Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
$523.00 - $1592.96
Catalog Number | Mfr. No. | Type | For Use With (Equipment) | Holds | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | For Use With (Equipment) | Holds | Price | Quantity | |||
11-679-084
|
Thermo Scientific™
185089 |
Canister Rack | All CryoMed controlled-rate freezers | Adjustable, 10-position |
Each for $1,592.96
|
|
|||
15-467-421
|
Thermo Scientific™
4000294 |
Straw Rack | All CryoMed controlled-rate freezers | 40 0.25/0.5 mL Straws |
Each for $523.00
|
|
|||
TF4000340
|
Thermo Scientific™
4000340 |
Canister Rack | All CryoMed controlled-rate freezers | 10 Canisters (for cord blood, Model 4000610) |
Each for $1,212.34
|
|
|||
15-467-461
|
Thermo Scientific™
4000701 |
Vial Rack (Small) | All CryoMed controlled-rate freezers | 76 1.2 mL/2 mL Ampules |
Each for $661.67
|
|
|||
15-467-462
|
Thermo Scientific™
4000702 |
Vial Rack Holder | All CryoMed controlled-rate freezers | Five 1.2 mL/2 mL Cryo Racks (Small) |
Each for $807.33
|
|
|||
15-467-463
|
Thermo Scientific™
4000703 |
Vial Rack (Large) | CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458, | 161 1.2 mL/2 mL Ampules |
Each for $789.78
|
|
|||
15-467-464
|
Thermo Scientific™
4000704 |
Vial Rack Holder | CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458 | Five 1.2 mL/2 mL Cryo Racks (Large) |
Each for $788.18
|
|
|||
13-262-837
|
Thermo Scientific™
4000705 |
Vial Rack (Small) | All CryoMed controlled-rate freezers | 76 4 mL/5 mL Ampules |
Each for $658.93
|
|
|||
13-262-838
|
Thermo Scientific™
4000706 |
Vial Rack Holder | All CryoMed controlled-rate freezers | Three 4 mL/5 mL Cryo Racks (Small) |
Each for $861.58
|
|
|||
13-262-839
|
Thermo Scientific™
4000707 |
Vial Rack (Large) | CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458 | 161 4 mL/5 mL Ampules |
Each for $764.24
|
|
|||
13-262-840
|
Thermo Scientific™
4000708 |
Vial Rack Holder | CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458 | Three 4 mL/5 mL Cryo Racks (Large) |
Each for $950.91
|
|
|||
Description
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
- Warranty varies by country, please contact us for details