missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BIOTIX™ Rainin LTS Compatible CleanPak Reload
BIOTIX™ Rainin LTS Compatible CleanPak Reload creates an ergonomic system in the lab with FlexFit™, StarStop™ and an LTS™ style pipette. Green Reload solution
$81.85 - $91.74
Specifications
Filtered | Non-filtered |
---|---|
Color | Clear |
Autoclavable | Autoclavable |
Certifications/Compliance | BSE/TSE free, Endotoxins/Pyrogen free, DNASE/RNASE free, PCR inhibitor free |
For Use With (Application) | Manual Volume Transfer |
Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Price | Quantity | ||||
12111376
|
BIOTIX™
63300010 |
20 μL | Sterile |
Pack of 960 Each for $82.06
Case of 5 Pack for $410.30
|
|
||||
12111377
|
BIOTIX™
63300011 |
250 μL | Sterile |
Pack of 960 Each for $82.06
Case of 5 Pack for $410.30
|
|
||||
12111378
|
BIOTIX™
63300012 |
300 μL | Sterile |
Pack of 960 Each for $87.38
Case of 5 Pack for $436.90
|
|
||||
12111379
|
BIOTIX™
63300013 |
1000 μL | Sterile |
Pack of 768 Each for $87.41
Case of 5 Pack for $437.05
|
|
||||
12111380
|
BIOTIX™
63300014 |
1200 μL | Sterile |
Pack of 768 Each for $82.06
Case of 5 Pack for $410.30
|
|
||||
12111381
|
BIOTIX™
63300025 |
20 μL | Non-sterile |
Pack of 960 Each for $91.74
Case of 5 Pack for $458.70
|
|
||||
12111382
|
BIOTIX™
63300026 |
250 μL | Non-sterile |
Pack of 960 Each for $81.88
Case of 5 Pack for $409.40
|
|
||||
12111383
|
BIOTIX™
63300027 |
300 μL | Non-sterile |
Pack of 960 Each for $81.88
Case of 5 Pack for $409.40
|
|
||||
12111384
|
BIOTIX™
63300028 |
1000 μL | Non-sterile |
Pack of 768 Each for $81.85
Case of 5 Pack for $409.25
|
|
||||
12111385
|
BIOTIX™
63300029 |
1200 μL | Non-sterile |
Pack of 768 Each for $81.85
Case of 5 Pack for $409.25
|
|
||||
Description
- FlexFit™ technology creates a seal with less insertion forces than that of competitors
- StarStop™ works with the Rainin LTS pipette to eliminate high ejection forces
- X-Resin™ maximizes sample recovery and accuracy by reducing surface to sample binding
- Blade™ reduces the frequency of hanging droplet formation, improving precision and accuracy
- CleanPak is fully recyclable and contains no contaminating cardboard
- Easily transfer snapcards to empty racks one handed
Order Info
960 tips/pack
Certifications
ISO 9001:2015, Certified free of DNASE, RNASE, Endotoxins/pyrogens, PCR inhibitors. Sterilized product is certified to 10^-3 SAL.
Specifications
Non-filtered | |
Autoclavable | |
Manual Volume Transfer | |
Ergonomic | |
Rainin LTS |
Clear | |
BSE/TSE free, Endotoxins/Pyrogen free, DNASE/RNASE free, PCR inhibitor free | |
FlexFit, StarStop, X-Resin, Blade | |
Single-use | |
Polypropylene |