RAM Scientific Safe-T-Fill Blood Gas Capillary Tube Mixing Components
Mixing flea and magnet for preventing clotting in blood gas capillary tube samples
Supplier:  RAM Scientific 079503
 View more versions of this product
Catalog No. 14-915-79
Explore available promotions
Description
Description
Magnetic rings and mixing fleas (mixing components) for SAFE-T-FILL™ Plastic Capillary Blood Gas Tubes prevent clotting in blood gas capillary tube samples.
  • Magnetic Rings measure Ø 8.8mm (width) x 3.9mm (length) ; hole diameter Ø 3.9mm
  • Mixing Fleas measure Ø 0.85mm (width) x 12mm (length)
  • Packaging labels are marked with Lot Number
Specifications
Specifications
Tube Mixing Fleas
Non-sterile
Intended to incorporate the blood sample with the anticoagulant by moving the mixing wire from side to side with the magnetic ring.
Stainless Steel
Capillary Blood Collection Tubes