RAM Scientific Safe-T-Fill™ Capillary Blood Collection Systems: Lithium Heparin
No scooping needed, allowing for high-quality, tissue-free samples
Supplier:  RAM Scientific 076101
 View more versions of this product
Catalog No. 14-915-64
Explore available promotions
Includes: Pre-assembled capillary blood collection device with a capillary tube, attached pierceable cap, and a microtube
Description
Description
No scooping needed, allowing for high-quality, tissue-free samples
  • 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
  • Blood samples are drawn into the microtube through capillary action, reducing hemolysis, tissue contamination, and re-draws. No "scooping" or "milking" required
  • Capillary straw and microtube are coated with anticoagulant Lithium Heparin (Green cap) to prevent clotting
  • Attached cap is color-coded to designate the type of additive that is in both the capillary tube and the microtube
  • Lot Number, Expiration Date, and fill lines are printed directly on the microtube
  • A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
  • For plasma determinations in chemistry
  • Small sample sizes, great for pediatrics and neonates
  • Optional gel separator serves as a barrier between blood and serum after centrifugation
  • Available in a variety of sizes and draw volumes, with or without gel separator
  • Optional amber microtube ensures light-sensitive sample integrity providing protection for testing (ex. Bilirubin)
Recommended Storage
Store at room temperature. Keep dry. Keep away from sunlight.
Specifications
Specifications
Clear
Lithium Heparin Mini
Non-sterile
125 μL
Green
Chemistry/Plasma/Whole Blood Tube
Plastic
Standard Pierceable Cap
Yes
Intended for chemistry determinations that require plasma or whole blood samples.