missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Raney™-Nickel, Activated Catalyst, 50% slurry in water, Thermo Scientific Chemicals

$193.66 - $193.66

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC395921000
SDS View Documents
Thermo Scientific Chemicals
395921000
100 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-02-0
58.69
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137 MFCD06798735
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]

Specifications

7440-02-0,7732-18-5,7429-90-5
MFCD00011137 MFCD06798735
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
[Ni]
58.69
CHEBI:28112
Plastic bottle
Raney™-Nickel, Activated Catalyst
Ni
100 g
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
935
58.69
Slurry
Safety and Handling

Safety and Handling

EINECSNumber : 231-111-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only