Reichert™ PG-Chek Digital Handheld Refractometer
Meets ASTM D3321-12 Standard test method for field test determination for freezing point of aqueous engine coolants
Supplier:  Reichert™ 13940026
 View more versions of this product
Catalog No. 13-940-026
Explore available promotions
Includes: 2 AAA Batteries
Description
Description
  • For precise measurement of Propylene Glycol coolant freeze point, boiling point and concentration percentage
  • Measuring scales: Fahrenheit and celsius
  • Automatic temperature compensation provides immediate, accurate, direct readings at ASTM D3321 standard
  • With IP65 dustproof/water resistant rating
  • Available with detachable neck lanyard and rubber side grips for easy handling
  • For 10000 readings
  • Available with auto-off sleep mode
Certifications
CE marked
Compliance
RoHS and WEEE
Warranty
One year (factory)
Specifications
Specifications
Distilled Water
589nm LED
IP65, RoHS, WEEE, CE
100 g
1°F Freezing point, 0.2%
2 AAA Batteries
3.5 oz.
68°F