missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Remco™ Polypropylene Hand Scoops

Molded one-piece design — no seams or cracks for bacterial growth

$18.29 - $19.29

Specifications

Product Type Hand Scoop
Material Polypropylene
Color Red
Certifications/Compliance FDA
For Use With (Application) Ideal for use where cross contamination is a concern
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Dimensions (L x W x H) Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Dimensions (L x W x H) Size Price Quantity  
29-635-554
View Documents
Remco™
64004
946.3 mL 11.5 x 5 x 2.75 in. (29.21 x 12.7 x 6.98 cm) Small
Each for $18.29
 
29-635-559
View Documents
Remco™
REM65004
2.42 L 15 x 6.5 x 3.5 in. (38.1 x 16.51 x 8.89 cm) Large
Each for $19.29
 
Description

Description

  • Made of thick-walled, FDA-compliant polypropylene
  • Ideal for use where cross-contamination is a concern
  • Chemical and corrosion resistant
  • Resilient and lightweight for easy handling
  • Withstand temperatures to 80°C (176°F)
  • Color: red

Specifications

Specifications

Hand Scoop
Red
Ideal for use where cross contamination is a concern
80°C
Polypropylene
FDA
176°F
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'