Restek™ Convenience Kits with Vial, Cap, and Septa: SilCote Deactivated DFS Item
Available with or without white graduated marking spots
Supplier:  Restek™ RK24674
 View more versions of this product
Catalog No. 06719007
Explore available promotions
Description
Description
  • Vials packaged in a clear-lid tray
  • Caps with septa packaged in a plastic bag
Specifications
Specifications
Amber
Silver
1000 Pk.
2 mL
Crimp Cap
11 mm