Restek™ Corporation Vu2 Union™ Connector Kit Connectors DFS Item
Vu2 Union connector combines the simplicity of a Press-Tight union with the strength of a metal union.
Supplier:  Restek™ RK21106
 View more versions of this product
Catalog No. 06-711-921
Description
Description
  • Connect a guard column to an analytical column
  • Connect a column to a transfer line
  • Connect two columns in series
  • Repair a broken column
  • Fit both Restek cage designs
Specifications
Specifications
Connector
0.28 to 0.32 mm
Vu2 Union body, two knurled nuts, two Press-Tight unions, and four ferrules
Vu2 Union Connector Kit